missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISOC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27082-1-AP-150UL
This item is not returnable.
View return policy
Description
ISOC2 gene ontology annotations related to this gene include protein destabilization.Specifications
| ISOC2 | |
| Polyclonal | |
| Unconjugated | |
| ISOC2 | |
| ISOC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 664994, 67441, 79763 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P85094, Q96AB3, Q9DCC7 | |
| ISOC2, Isoc2a, Isoc2b | |
| ISOC2 Fusion Protein Ag23936 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion