Learn More
ISOC1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25844-1-AP-150UL
 View more versions of this product
Product Code. 17281376
  • £349.00 / 150µL
Description
The ISOC1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ISOC1. This antibody recognizes human, mouse antigen. The ISOC1 antibody has been validated for the following applications: IP, WB,ELISA analysis.
Specifications
Specifications
ISOC1
Polyclonal
Unconjugated
150 μL
51015
Liquid
Western Blot, Immunoprecipitation, ELISA
300 μg/mL
Rabbit
Primary
Human, Mouse