missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl Chloride, TRC

Product Code. 30392747
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392747 Supplier Toronto Research Chemicals Supplier No. TRCI7806805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutyryl Chloride
CAS 79-30-1
Molecular Formula C4H7ClO
Quantity 5 g
Synonym 2-Methylpropanoyl Chloride, 2,2-Dimethylacetyl Chloride, 2-Methylpropanoic Acid Chloride, 2-Methylpropanoyl Chloride, 2-Methylpropionyl Chloride, 2-Propylcarbonyl Chloride, Chloro Isopropyl Ketone, Dimethylacetyl Chloride, Isobutanoyl Chloride, Isobutyric Acid Chloride, Isobutyroyl Chloride, Isopropanecarbonyl Chloride, i-Butyryl Chloride, alpha-Methylpropionyl Chloride
InChI Formula InChI=1S/C4H7ClO/c1-3(2)4(5)6/h3H,1-2H3
SMILES CC(C)C(=O)Cl
IUPAC Name 2-methylpropanoyl chloride
Molecular Weight (g/mol) 106.55
Formula Weight 106.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.