missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl Carnitine D3, TRC

Product Code. 30395418
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395418 10 mg 10mg
30395302 5 mg 5mg
30391915 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395418 Supplier Toronto Research Chemicals Supplier No. TRCI77902710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutryl Carnitine D3
Molecular Formula C11 D3 H18 N O4
Quantity 10 mg
InChI Formula InChI=1S/C11H21NO4/c1-8(2)11(15)16-9(6-10(13)14)7-12(3,4)5/h8-9H,6-7H2,1-5H3/t9-/m1/s1/i1D3/t8?,9-
SMILES [2H]C([2H])([2H])C(C)C(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C
IUPAC Name (3R)-3-(3,3,3-trideuterio-2-methylpropanoyl)oxy-4-(trimethylazaniumyl)butanoate
Molecular Weight (g/mol) 234.307
Formula Weight 234.166
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.