missing translation for 'onlineSavingsMsg'
Learn More

Isobutyrophenone, 97%

Product Code. 11400498
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11400498

Brand: Thermo Scientific Alfa Aesar B24628.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-70-1 | C10H12O | 148.21 g/mol

Isobutyrophenone is used as an internal standard in the determination of hydroxyzine hydrochloride and benzyl alcohol in injection solutions. It is also used in the preparation of alpha-hydroxyisobutyrophenone. Isobutyrophenone is employed as photosensitizer intermediate. It can react with bromoacetic acid ethyl ester to get 3-hydroxy-4-methyl-3-phenyl-valeric acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyrophenone is used as an internal standard in the determination of hydroxyzine hydrochloride and benzyl alcohol in injection solutions. It is also used in the preparation of alpha-hydroxyisobutyrophenone. Isobutyrophenone is employed as photosensitizer intermediate. It can react with bromoacetic acid ethyl ester to get 3-hydroxy-4-methyl-3-phenyl-valeric acid ethyl ester.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-70-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008917
InChI Key BSMGLVDZZMBWQB-UHFFFAOYSA-N
Synonym isobutyrophenone, isopropyl phenyl ketone, 2-methylpropiophenone, 1-propanone, 2-methyl-1-phenyl, phenyl isopropyl ketone, isopropyl phenyl keton, alpha-methylpropiophenone, unii-8l53972ntz, .alpha.-methylpropiophenone, 2-methyl-1-phenyl-1-propanone
PubChem CID 69144
IUPAC Name 2-methyl-1-phenylpropan-1-one
SMILES CC(C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 1°C
Density 0.988
Boiling Point 217°C
Flash Point 84°C (184°F)
Refractive Index 1.517
Quantity 25 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material Isobutyrophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.