missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%

Product Code. 11393526
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393526 250 mL 250mL
11303536 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393526 Supplier Thermo Scientific Alfa Aesar Supplier No. L14470.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-82-0 | C4H7N | 69.107 g/mol

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001873
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
PubChem CID 6559
ChEBI CHEBI:28638
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

Melting Point -72°C
Density 0.77
Boiling Point 103°C to 104°C
Flash Point 8°C (46°F)
Linear Formula (CH3)2CHCN
Refractive Index 1.374
Quantity 250 mL
UN Number UN2284
Beilstein 1340512
Merck Index 14,5156
Solubility Information Soluble in water 35g/L (20°C).
Formula Weight 69.11
Percent Purity 99%
Chemical Name or Material Isobutyronitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyronitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Isobutyronitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.