missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%

Product Code. 10548850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1L; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10548850 1L; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10548850 Supplier Thermo Scientific Acros Supplier No. 122530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-82-0 | C4H7N | 69.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-82-0
Melting Point -72°C
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 107°C to 108°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H7N
Linear Formula (CH3)2CHCN
Refractive Index 1.3720 to 1.374
MDL Number MFCD00001873
Quantity 1 L
Beilstein 02, 294
Merck Index 15, 5201
Specific Gravity 0.76
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
Solubility Information Solubility in water: 35g/L (20 c). Other solubilities: soluble in alcohol and ether
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
SMILES CC(C)C#N
IUPAC Name 2-methylpropanenitrile
Molecular Weight (g/mol) 69.11
PubChem CID 6559
ChEBI CHEBI:28638
Formula Weight 69.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isobutyronitrile, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyronitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.