missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyric acid-benzyl ester, Dr. Ehrenstorfer
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Isobutyric acid-benzyl ester |
| CAS | 103-28-6 |
| Analyte or Component Names | Isobutyric acid-benzyl ester |
| Shipping Condition | +20°C |
| Molecular Formula | C11 H14 O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Isobutyric acid, benzyl ester (6CI,7CI,8CI), Benzyl 2-methylpropionate, Benzyl isobutanoate, Benzyl isobutyrate, NSC 406201, NSC 46112, Phenylmethyl 2-methylpropanoate |
| InChI Formula | InChI=1S/C11H14O2/c1-9(2)11(12)13-8-10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3 |
| SMILES | CC(C)C(=O)OCc1ccccc1 |
| IUPAC Name | benzyl 2-methylpropanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction