missing translation for 'onlineSavingsMsg'
Learn More

Isobutyramide, 99%

Product Code. 11480197
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11480197

Brand: Thermo Scientific Alfa Aesar B21641.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 563-83-7 | C4H9NO | 87.122 g/mol

Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

Solubility
Soluble in water.

Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-83-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00008019
InChI Key WFKAJVHLWXSISD-UHFFFAOYSA-N
Synonym isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
PubChem CID 68424
IUPAC Name 2-methylpropanamide
SMILES CC(C)C(=O)N

Specifications

Melting Point 127°C to 129°C
Density 1.013
Boiling Point 216°C to 220°C
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material Isobutyramide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.