missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde, 98%

Product Code. 11420033
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420033 2500 mL 2500mL
11410033 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11420033 Supplier Thermo Scientific Alfa Aesar Supplier No. A12106.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 78-84-2 | C4H8O | 72.11 g/mol

Isobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-84-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00006980
InChI Key AMIMRNSIRUDHCM-UHFFFAOYSA-N
Synonym isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
PubChem CID 6561
ChEBI CHEBI:48943
IUPAC Name 2-methylpropanal
SMILES CC(C)C=O

Specifications

Melting Point -65°C
Density 0.794
Boiling Point 63°C to 65°C
Flash Point −24°C (−11°F)
Assay Percent Range 98%
Odor Pungent
Linear Formula (CH3)2CHCHO
Refractive Index 1.3723
Quantity 2500 mL
UN Number UN2045
Beilstein 605330
Sensitivity Air sensitive
Merck Index 14,5154
Solubility Information Slightly miscible with water.
Formula Weight 72.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.