missing translation for 'onlineSavingsMsg'
Learn More

Isobutyltriethoxysilane, 95%, Thermo Scientific™

Product Code. 15495788
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495788 5 g 5g
15405798 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495788 Supplier Thermo Scientific Alfa Aesar Supplier No. H59461.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in a wide range of applications including silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent, silane cross linking agent, silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent, silane reinforcer, silylating agent, silane reducing agent, silyl building blocks and synthons.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 17980-47-1
Molecular Formula C10H24O3Si
Molecular Weight (g/mol) 220.384
MDL Number MFCD00069084
InChI Key ALVYUZIFSCKIFP-UHFFFAOYSA-N
Synonym triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilane
PubChem CID 87391
IUPAC Name triethoxy(2-methylpropyl)silane
SMILES CCO[Si](CC(C)C)(OCC)OCC

Spécification

Boiling Point 165°C (400mmHg)
Melting Point 190°C to 191°C
Physical Form Liquid
Quantity 5 g
Linear Formula (CH3)2CHCH2Si(OC2H5)3
Beilstein 1750049
Solubility Information Insoluble in water.
Formula Weight 220.38
Percent Purity 95%
Refractive Index 1.4505
Density 0.880 g/mL
Chemical Name or Material Isobutyltriethoxysilane
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.