missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene oxide, 99+%

Product Code. 11372257
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372257 5 g 5g
11392257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372257 Supplier Thermo Scientific Alfa Aesar Supplier No. L09773.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 558-30-5 | C4H8O | 72.107 g/mol

Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.

Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-30-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00066354
InChI Key GELKGHVAFRCJNA-UHFFFAOYSA-N
Synonym isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
PubChem CID 11208
IUPAC Name 2,2-dimethyloxirane
SMILES CC1(CO1)C

Specifications

Density 0.808
Boiling Point 51°C to 52°C
Flash Point −32°C (−25°F)
Odor Ether-like
Refractive Index 1.375
Quantity 5 g
UN Number UN2924
Beilstein 102408
Solubility Information Miscible with ethanol and diethyl ether. Slightly miscible with water.
Formula Weight 72.11
Percent Purity ≥99%
Chemical Name or Material Isobutylene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.