missing translation for 'onlineSavingsMsg'
Learn More

Isobutylboronic acid, 97%

Product Code. 11372799
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372799 5 g 5g
11362799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372799 Supplier Thermo Scientific Alfa Aesar Supplier No. H27698.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84110-40-7 | C4H11BO2 | 101.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84110-40-7
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00134156
InChI Key ZAZPDOYUCVFPOI-UHFFFAOYSA-N
Synonym isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid
PubChem CID 2734395
IUPAC Name 2-methylpropylboronic acid
SMILES CC(C)CB(O)O

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Formula Weight 101.94
Percent Purity 97%
Chemical Name or Material Isobutylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.