missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl vinyl ether 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Isobutyl Vinyl Ether |
| CAS | 109-53-5 |
| Analyte or Component Names | Isobutyl Vinyl Ether |
| Shipping Condition | +20°C |
| Molecular Formula | C6 H12 O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ether, isobutyl vinyl (8CI), 1-(Ethenyloxy)-2-methylpropane, IVE, Isobutanol vinyl ether, Isobutoxyethene, Isobutyl vinyl ether, Lutonal LR 8500, NSC 8265, Vinoflex MO 400, Vinyl isobutyl ether, iso-Butyl vinyl ether, Propane, 1-(ethenyloxy)-2-methyl- |
| InChI Formula | InChI=1S/C6H12O/c1-4-7-5-6(2)3/h4,6H,1,5H2,2-3H3 |
| SMILES | CC(C)COC=C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?