missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Thiuram Disulfide, TRC

Product Code. 30392471
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392471 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392471 Supplier Toronto Research Chemicals Supplier No. TRCI78875050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutyl Thiuram Disulfide
CAS 3064-73-1
Molecular Formula C18 H36 N2 S4
Quantity 50 mg
InChI Formula InChI=1S/C18H36N2S4/c1-13(2)9-19(10-14(3)4)17(21)23-24-18(22)20(11-15(5)6)12-16(7)8/h13-16H,9-12H2,1-8H3
SMILES CC(C)CN(CC(C)C)C(=S)SSC(=S)N(CC(C)C)CC(C)C
IUPAC Name bis(2-methylpropyl)carbamothioylsulfanyl N,N-bis(2-methylpropyl)carbamodithioate
Molecular Weight (g/mol) 408.752
Formula Weight 408.176
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.