missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Sildenafil, TRC

Product Code. 30395415
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395415 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395415 Supplier Toronto Research Chemicals Supplier No. TRCI78065050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[2-Ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-(2-methylpropyl)-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
CAS 1391053-95-4
Molecular Formula C23 H32 N6 O4 S
Quantity 50 mg
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-(2-methylpropyl)-, 5-[2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-(2-methylpropyl)-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Sildenafil Citrate Imp. A (EP), Sildenafil Related Compound A (USP)
InChI Formula InChI=1S/C23H32N6O4S/c1-6-33-19-8-7-16(34(31,32)29-11-9-27(4)10-12-29)14-17(19)22-24-20-18(13-15(2)3)26-28(5)21(20)23(30)25-22/h7-8,14-15H,6,9-13H2,1-5H3,(H,24,25,30)
SMILES CCOc1ccc(cc1C2=Nc3c(CC(C)C)nn(C)c3C(=O)N2)S(=O)(=O)N4CCN(C)CC4
IUPAC Name 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-(2-methylpropyl)-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol) 488.6
Formula Weight 488.2206
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.