missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl p-toluenesulfonate, 97%

Product Code. 11480571
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480571 25 g 25g
11470571 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480571 Supplier Thermo Scientific Alfa Aesar Supplier No. B20080.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4873-56-7 | C11H16O3S | 228.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4873-56-7
Molecular Formula C11H16O3S
Molecular Weight (g/mol) 228.306
MDL Number MFCD00026478
InChI Key QIMRPGAQCKHRKB-UHFFFAOYSA-N
Synonym isobutyl p-toluenesulfonate, p-toluenesulfonic acid isobutyl ester, isopropyl 4-toluene sulphonate, isobutyl 4-methylbenzenesulfonate, 2-methylpropyl 4-methylbenzene-1-sulfonate, benzenesulfonic acid, 4-methyl-, 2-methylpropyl ester, ccris 9147, isobutyl tosylate, p-toluenesulfonic acid, isobutyl ester, isobutylp-toluenesulfonate
PubChem CID 310590
IUPAC Name 2-methylpropyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(C)C

Specifications

Refractive Index 1.5025
Quantity 25 g
Formula Weight 228.28
Percent Purity 97%
Chemical Name or Material Isobutyl p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.