missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutyl nitrite, 95%, stab. with potassium carbonate

Product Code. 15468829
Change view
Click to view available options
Quantity:
25mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468829 25mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15468829 Supplier Alfa Aesar™ Supplier No. L05259.AC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 542-56-3
Boiling Point 66°C to 67°C
Flash Point −21°C (−5°F)
Molecular Formula C4H9NO2
Linear Formula (CH3)2CHCH2ONO
MDL Number MFCD00002056
Quantity 25mL
UN Number UN2351
Beilstein 1699518
Merck Index 14,5149
Synonym isobutyl nitrite, isobutylnitrite, nitrous acid, 2-methylpropyl ester, nitrous acid, isobutyl ester, blackjack, isobutylnitrit, unii-gw9wab6qom, ccris 1099, ibn, nitrous acid 2-methylpropyl ester
InChI Key APNSGVMLAYLYCT-UHFFFAOYSA-N
SMILES CC(C)CON=O
IUPAC Name 2-methylpropyl nitrite
Molecular Weight (g/mol) 103.121
PubChem CID 10958
ChEBI CHEBI:46643
Formula Weight 103.12
Percent Purity 95%
Chemical Name or Material Isobutyl nitrite, stabilized with potassium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.