missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99%, ACROS Organics™

Brand:  Acros Organics 412680010

Code : 29

Additional Details : CAS Number : 97-86-9 Weight : 1.05000kg Transport : UN number : 2283 Chem class : 1 Pack group : III

Quantity 1kg
Packaging Glass Bottle

 View more versions of this product

Product Code. 10239460

  • £53.40 / 1kg


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

Isobutyl methacrylate
97-86-9
99%
CH2=C(CH3)COOCH2CH(CH3)2
MFCD00008931
Stabilized
99%
C8H14O2
142.2

Specifications

Isobutyl methacrylate
1kg
97-86-9
0.8800g/mL
99%
C8H14O2
142.2
Authentic
1.4190 to 1.4210
0.1% max.
Clear colorless
46°C
0.88
Stabilized
0.005% max. (as methacrylic acid)
Liquid
99%
CH2=C(CH3)COOCH2CH(CH3)2
MFCD00008931
Glass Bottle
15 to 20ppm MEHQ
155.0°C
− 37.0°C
Videos
SDS