missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Isobutyrate 99%, ACROS Organics™

Brand:  Acros Organics 412675000

Code : 29

Additional Details : CAS Number : 97-85-8 Weight : 0.55000kg Transport : UN number : 2528 Chem class : 1 Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10005810

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Isobutyl isobutyrate
97-85-8
99%
(CH3)2CHCOOCH2CH(CH3)2
MFCD00008916
99%
99%
C8H16O2
144.21
IBIB

Specifications

Isobutyl isobutyrate
500g
97-85-8
Liquid
99%
(CH3)2CHCOOCH2CH(CH3)2
MFCD00008916
Authentic
Glass bottle
147.0° to 149.0°C (1013.0 hPa)
−81.0°C
0.856
99%
0.8560g/mL
99%
C8H16O2
144.21
15, 5190
IBIB
1.3980 to 1.4000
Clear
38°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isobutyl isobutyrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS