missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Ethyl Ether, TRC

Product Code. 30399451
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399451 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399451 Supplier Toronto Research Chemicals Supplier No. TRCI6270501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Ethoxy-2-methylpropane
CAS 627-02-1
Molecular Formula C6 H14 O
Quantity 1 g
Synonym Isobutyl Ethyl Ether, Propane, 1-ethoxy-2-methyl-, Ether, ethyl isobutyl (6CI,7CI,8CI), 1-Ethoxy-2-methylpropane, Ethyl iso-butyl ether, Ethyl isobutyl ether, Isobutyl ethyl ether
InChI Formula InChI=1S/C6H14O/c1-4-7-5-6(2)3/h6H,4-5H2,1-3H3
SMILES CCOCC(C)C
IUPAC Name 1-ethoxy-2-methylpropane
Molecular Weight (g/mol) 102.175
Formula Weight 102.104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.