missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl chloroformate, 98%

Product Code. 11457696
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457696 500 g 500g
11437696 25 g 25g
11447696 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11457696 Supplier Thermo Scientific Alfa Aesar Supplier No. A14692.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 543-27-1 | C5H9ClO2 | 136.575 g/mol

Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-27-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00000642
InChI Key YOETUEMZNOLGDB-UHFFFAOYSA-N
Synonym isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
PubChem CID 62365
IUPAC Name 2-methylpropyl carbonochloridate
SMILES CC(C)COC(=O)Cl

Specifications

Melting Point -80°C
Density 1.048
Boiling Point 128°C to 129°C
Flash Point 30°C (86°F)
Assay Percent Range 98%
Linear Formula ClCO2CH2CH(CH3)2
Refractive Index 1.406
Quantity 500 g
UN Number UN2742
Beilstein 956590
Sensitivity Moisture sensitive
Merck Index 14,5139
Solubility Information Immiscible with water.
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.