missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl chloroformate, 98%, AcroSeal™

Product Code. 10155777 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155777 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10155777 Supplier Thermo Scientific Acros Supplier No. 430811000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 543-27-1 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

Acidity 0.1% max.
CAS 543-27-1
Density 1.0475g/mL
Boiling Point 128.7°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. isobutyl carbonate (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C5H9ClO2
Linear Formula ClCO2CH2CH(CH3)2
Refractive Index 1.4060 to 1.4080
MDL Number MFCD00000642
Quantity 100 mL
Merck Index 15,5184
Specific Gravity 1.0475
Synonym isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes in alcohol,miscible with benzene,chloroform,acetone,toluene
InChI Key YOETUEMZNOLGDB-UHFFFAOYSA-N
SMILES CC(C)COC(=O)Cl
IUPAC Name 2-methylpropyl carbonochloridate
Molecular Weight (g/mol) 136.58
PubChem CID 62365
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.