missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol

Product Code. 11323346
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323346 500 mL 500mL
11313346 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323346 Supplier Thermo Scientific Alfa Aesar Supplier No. L12739.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-63-8 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-63-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00042865
InChI Key CFVWNXQPGQOHRJ-UHFFFAOYSA-N
Synonym isobutyl acrylate, isobutyl propenoate, isobutyl 2-propenoate, 2-propenoic acid, 2-methylpropyl ester, acrylic acid, isobutyl ester, 2-methylpropyl acrylate, isobutylester kyseliny akrylove, propenoic acid, isobutyl ester, ccris 4828, hsdb 610
PubChem CID 7819
IUPAC Name 2-methylpropyl prop-2-enoate
SMILES CC(C)COC(=O)C=C

Specifications

Melting Point -61°C
Density 0.888
Boiling Point 139°C
Flash Point 32°C (89°F)
Linear Formula CH2=CHCO2CH2CH(CH3)2
Refractive Index 1.415
Quantity 500 mL
UN Number UN2527
Beilstein 1749388
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material Isobutyl acrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.