missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Acetate, 99%, Honeywell™

Product Code. p-7120497 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15638420 25 mL Glass Bottle 25mL
15605780 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15638420 Supplier Honeywell Research Chemicals Supplier No. 53747025ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Melting Point -99°C
Color Colorless
Density 0.867g/mL at 25°C
Boiling Point 117°C
Explosion Limit 0.105
Flash Point 22°C (71.6°F)
Ignition Point 793°F
Packaging Glass Bottle
Linear Formula CH3COOCH2CH(CH3)2
Refractive Index n20/D 1.39
Quantity 25 mL
UN Number UN1213
Beilstein 1741909
Vapor Density >4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (20°C)
Percent Purity 99%
Chemical Name or Material Isobutyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.