missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acetate, 98%

Product Code. 11412697
Change view
Click to view available options
Quantity:
250 mL
500 mL
2500 mL
Unit Size:
2500mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412697 2500 mL 2500mL
11468870 250 mL 250mL
11402697 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412697 Supplier Thermo Scientific Alfa Aesar Supplier No. A14892.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-19-0 | C6H12O2 | 116.16 g/mol

Isobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.

Solubility
Miscible with ethanol, ether, methanol and acetone. Slightly miscible with water and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Melting Point -99°C
Density 0.87
Boiling Point 116°C to 117°C
Flash Point 18°C (64°F)
Assay Percent Range 98%
Odor Fruit-like
Refractive Index 1.3895
Quantity 2500 mL
UN Number UN1213
Beilstein 1741909
Merck Index 14,5130
Solubility Information Miscible with ethanol,ether,methanol and acetone. Slightly miscible with water and carbon tetrachloride.
Formula Weight 116.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Isobutyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.