missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl 4-hydroxybenzoate, 98%

Product Code. p-7047094
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318536 5 g 5g
11328536 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318536 Supplier Thermo Scientific Alfa Aesar Supplier No. L17587.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4247-02-3 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-4247
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020167
InChI Key XPJVKCRENWUEJH-UHFFFAOYSA-N
Synonym isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
PubChem CID 20240
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
SMILES CC(C)COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
Beilstein 2642305
Solubility Information Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Isobutyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.