missing translation for 'onlineSavingsMsg'
Learn More

Isobutanol, ACS, 99+%

Product Code. 11398068
Change view
Click to view available options
Quantity:
500 mL
1 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398068 1 L 1 L
11495180 500 mL 500mL
11318078 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398068 Supplier Thermo Scientific Alfa Aesar Supplier No. 036643.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.001%, Solubility in water P.T., Titratable acid 0.0005meq/g, H2O 0.1%, Carbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone) | CAS: 78-83-1 | C4H10O | 74.123 g/mol

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point °C to 108°C
Color Colorless
Density 0.802 g/mL
Quantity 1 L
Flash Point 28°C (82°F)
Physical Form Liquid
Refractive Index 1.396
Assay Percent Range >99+%
Beilstein 1730878
Boiling Point 107°C to 108°C
Solubility Information Miscible with alcohol and diethyl ether. Partially miscible with water.
Formula Weight 74.12
Grade ACS
Chemical Name or Material Isobutanol, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Isobutyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.