Learn More
Isobutanol, HPLC Grade, 99+%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%
£60.20 - £206.00
Chemical Identifiers
CAS | 78-83-1 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00004740 |
InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
Synonym | 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol |
PubChem CID | 6560 |
ChEBI | CHEBI:46645 |
IUPAC Name | 2-methylpropan-1-ol |
SMILES | CC(C)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388147
|
Thermo Scientific Alfa Aesar
022908.K2 |
1 L |
£60.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398147
|
Thermo Scientific Alfa Aesar
022908.M1 |
4 x 1 L |
£206.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308157
|
Thermo Scientific Alfa Aesar
022908.K7 |
4 L |
£163.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-83-1 | |
74.123 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Specifications
78-83-1 | |
Colorless | |
107°C to 108°C | |
390°C (734°F) | |
Sweet | |
1.396 | |
1212 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
74.12 | |
HPLC | |
Isobutanol |
-108°C | |
0.802 g/cm3 | |
28°C (82°F) | |
>99% | |
C4H10O | |
MFCD00004740 | |
1730878 | |
Miscible with alcohol and diethyl ether. Partially miscible with water. | |
CC(C)CO | |
74.123 | |
CHEBI:46645 | |
4-40 hPa (3-30mm Hg) at 20°C | |
Liquid |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335-H336
missing translation for 'dotInformation' : Hazard Class: 3, Packing Group: III
missing translation for 'einecsNumber' : 201-148-0
missing translation for 'rtecsNumber' : NP9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only