missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutanol, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
2500 mL
Descripción
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
Solubility
Miscible with alcohol and diethyl ether. Partially miscible with water.
Notes
Avoid all sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
Solubility
Miscible with alcohol and diethyl ether. Partially miscible with water.
Notes
Avoid all sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Especificaciones
pH | 7 |
Melting Point | -108°C |
Density | 0.802 |
Boiling Point | 107°C to 108°C |
Quantity | 2500 mL |
Flash Point | 28°C (82°F) |
Odor | Sweet |
Refractive Index | 1.396 |
Linear Formula | (CH3)2CHCH2OH |
UN Number | UN1212 |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'