missing translation for 'onlineSavingsMsg'
Learn More

Isobutanamidoxime, TRC

Product Code. 30389408
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389408 Supplier Toronto Research Chemicals Supplier No. TRCI78910025G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutanamidoxime
CAS 35613-84-4
Molecular Formula C4H10N2O
Quantity 25 g
Synonym Isobutyramide Oxime, Isobutyramidoxime, Isopropylcarboxamide Oxime, N-Hydroxy-2-methylpropanimidamide, N-Hydroxyisobutyramidine, N'-Hydroxyisobutyramidine
InChI Formula InChI=1S/C4H10N2O/c1-3(2)4(5)6-7/h3,7H,1-2H3,(H2,5,6)
SMILES CC(C)\C(=N\O)\N
Molecular Weight (g/mol) 102.14
Formula Weight 102.0793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.