missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl acetate, 94%

Product Code. p-7045723
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11302607

Brand: Thermo Scientific Alfa Aesar L11748.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 125-12-2 | C12H20O2 | 196.29 g/mol

Isobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-12-2
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135943
InChI Key KGEKLUUHTZCSIP-JFGNBEQYSA-N
Synonym 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate
PubChem CID 6950273
IUPAC Name [(1S,3S,4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C

Specifications

Density 0.99
Boiling Point 220°C to 224°C
Flash Point 89°C (192°F)
Refractive Index 1.465
Quantity 100 mL
Beilstein 3197572
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 196.29
Percent Purity 94%
Chemical Name or Material Isobornyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.