missing translation for 'onlineSavingsMsg'
Learn More

Isoborneol, Dr. Ehrenstorfer

Product Code. 30325898
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325898 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325898 Supplier Dr. Ehrenstorfer Supplier No. DREC14384900
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoborneol
CAS 124-76-5
Analyte or Component Names Isoborneol
Shipping Condition Room Temperature
Molecular Formula C10 H18 O
Certifications/Compliance ISO/IEC 17025
Synonym Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1R,2R,4R)-rel-, Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, exo-, Isoborneol (8CI), rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol, (±)-Isoborneol, 2-exo-Bornyl alcohol, DL-Isoborneol, Isobornyl alcohol, NSC 26350, dl-Isoborneol, exo-2-Hydroxy-1,7,7-trimethylnorbornane
InChI Formula InChI=1S/C10H18O/c1-9(2)7-4-5-10(9,3)6-8(7)11/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1
SMILES C[C@@]12CC[C@@H]([C@H](O)C1)C2(C)C
IUPAC Name (1R,2R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol) 154.25
Quantity 100 mg
Formula Weight 154.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.