missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobavachalcone, MedChemExpress
Description
Isobavachalcone (Corylifolinin) is derived from Psoralea corylifolia Linn. and is a potent inhibitor of Akt signaling pathway, which induces apoptosis in human cancer cells (Inhibits OVCAR-8 cell growth with an IC50 value of 7.92 μM). Isobavachalcone also induces Reactive Oxyen Species (ROS) generation in OVCAR-8 cells and has exhibit cancer anti-promotive and anti-proliferative activity.
Specifications
Specifications
| Chemical Name or Material | Isobavachalcone |
| CAS | 20784-50-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H20O4 |
| Quantity | 10 mg |
| Synonym | Corylifolinin Isobacachalcone |
| Solubility Information | DMSO : 11 mg/mL (33.91 mM; Need ultrasonic and warming) |
| SMILES | O=C(/C=C/C1=CC=C(C=C1)O)C2=CC=C(C(C/C=C(C)\C)=C2O)O |
| Molecular Weight (g/mol) | 324.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?