missing translation for 'onlineSavingsMsg'
Learn More

Isoastragaloside II, MedChemExpress

Product Code. 16431578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431578 5 mg 5mg
16429168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431578 Supplier Medchem Express Supplier No. HYN0888
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isoastragaloside II is an astragaloside, which is isolated from the hairy root culture of Astragalus membranaceus.

Isoastragaloside II is an astragaloside, which is isolated from the hairy root culture of Astragalus membranaceus.

Specifications

Chemical Name or Material Isoastragaloside II
CAS 86764-11-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C43H70O15
Quantity 5 mg
Synonym Astrasieversianin-VII
Solubility Information DMSO : 100 mg/mL (120.92 mM; Need ultrasonic)
SMILES C[C@]([C@@]1(CC2)C)(C[C@H](O)[C@]1([H])[C@@]3(O[C@H](C(C)(O)C)CC3)C)[C@@](C[C@H](O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)[C@@]5([H])C6(C)C)([H])[C@@]72[C@]5(CC[C@@H]6O[C@@](OC[C@@H](O)[C@@H]8OC(C)=O)([H])[C@@H]8O)C7
Molecular Weight (g/mol) 827.01
Formula Weight 827.01
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.