missing translation for 'onlineSavingsMsg'
Learn More

Isoanastrozole Monoamide, TRC

Product Code. 30393381
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393381 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393381 Supplier Toronto Research Chemicals Supplier No. TRCI77997010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Isoanastrozole Monoamide
Analyte or Component Names Isoanastrozole Monoamide
Shipping Condition Room Temperature
Molecular Formula C17H21N5O
Synonym 2-(3-((4H-1,2,4-Triazol-4-yl)methyl)-5-(2-cyanopropan-2-yl)phenyl)-2-methylpropanamide
InChI Formula InChI=1S/C17H21N5O/c1-16(2,9-18)13-5-12(8-22-10-20-21-11-22)6-14(7-13)17(3,4)15(19)23/h5-7,10-11H,8H2,1-4H3,(H2,19,23)
Solution Type Neat
SMILES CC(C)(C#N)c1cc(Cn2cnnc2)cc(c1)C(C)(C)C(=O)N
Industry Type Pharmaceutical
Molecular Weight (g/mol) 311.38
Quantity 10 mg
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.