missing translation for 'onlineSavingsMsg'
Learn More

Isoamylamine, TRC

Product Code. 30402681
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402681 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402681 Supplier Toronto Research Chemicals Supplier No. TRCI7800055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107-85-7
Molecular Formula C5 H13 N
Quantity 5 g
Synonym 1-Butanamine, 3-methyl- (9CI, ACI), 3-Methyl-1-butanamine (ACI), Isopentylamine (6CI, 8CI), Propylamine, 3,3-dimethyl- (7CI), 1-Amino-3-methylbutane, 2-(2-Isopropyl)ethylamine, 3-Methyl-1-aminobutane, 3-Methyl-n-butylamine, 3-Methylbutanamine, 3-Methylbutylamine, Isoamylamine, Isovalerylamine, Leucamine, Monoisoamylamine, Monoisopentylamine, NSC 7907, γ-Isoamylamine
InChI Formula InChI=1S/C5H13N/c1-5(2)3-4-6/h5H,3-4,6H2,1-2H3
SMILES CC(C)CCN
IUPAC Name 3-methylbutan-1-amine
Molecular Weight (g/mol) 87.16
Formula Weight 87.1048
Chemical Name or Material 3-Methylbutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.