missing translation for 'onlineSavingsMsg'
Learn More

Isoamylamine, 98+%

Product Code. 10612983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10612983 5 mL Glass bottle 5mL
10449070 25 mL Glass bottle 25mL
10677124 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612983 Supplier Thermo Scientific Acros Supplier No. 155470050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-85-7 | C5H13N | 87.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-85-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.15
MDL Number MFCD00008203
InChI Key BMFVGAAISNGQNM-UHFFFAOYSA-N
Synonym isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
PubChem CID 7894
ChEBI CHEBI:43689
IUPAC Name 3-methylbutan-1-amine
SMILES CC(C)CCN

Specifications

Melting Point -60.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 95.0°C to 97.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2NH2
Refractive Index 1.4070 to 1.4100
Quantity 5 mL
UN Number 1106
Beilstein 04, 180
Merck Index 15, 5157
Specific Gravity 0.75
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 87.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isoamylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isoamylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.