missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Alcohol, TRC

Product Code. 30393201
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393201 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393201 Supplier Toronto Research Chemicals Supplier No. TRCI780010100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoamyl Alcohol (3-Methyl-1-Butanol)
CAS 123-51-3
Purity Grade Notes GC
Molecular Formula C5 H12 O
Quantity 100 g
Synonym 3-Methyl-1-butanol, 1-Butanol, 3-methyl-, Isopentyl alcohol (8CI), 3-Methyl-1-butanol, 2-Methyl-4-butanol, 3-Methylbutane-1-ol, 3-Methylbutyl alcohol, Isoamyl alcohol, Isoamylol, Isobutyl carbinol, Isopentanol, NSC 1029, NSC 7905, iso-Amyl alcohol, iso-Pentanol
InChI Formula InChI=1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3
SMILES CC(C)CCO
IUPAC Name 3-methylbutan-1-ol
Molecular Weight (g/mol) 88.15
Formula Weight 88.0888
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.