missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Acetate, TRC

Product Code. 30133015
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133015 50 mL 50mL
30137367 25 mL 25mL
30009317 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133015 Supplier Toronto Research Chemicals Supplier No. TRCI78002050ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7 H14 O2
Molecular Weight (g/mol) 130.18
Synonym 1-Butanol, 3-methyl-, acetate (9CI), Acetic acid, isoamyl ester (3CI), Isopentyl alcohol, acetate (6CI,8CI), 3-Methyl-1-butanol acetate, 3-Methyl-1-butyl acetate, 3-Methylbutyl acetate, 3-Methylbutyl ethanoate, Acetic acid 3-methyl-1-butyl ester, Acetic acid 3-methylbutyl ester, Acetic acid isopentyl ester, Banana oil, Isoamyl acetate, Isoamyl alcohol acetate, Isoamyl ethanoate, Isopentyl acetate, Isopentyl ethanoate, NSC 9260, Pear oil, i-Amyl acetate, iso-Amyl acetate, iso-Pentyl acetate
IUPAC Name 3-methylbutyl acetate
SMILES CC(C)CCOC(=O)C

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C7H14O2/c1-6(2)4-5-9-7(3)8/h6H,4-5H2,1-3H3
Formula Weight 130.0994
Percent Purity >95%
Chemical Name or Material Acetic acid-3-methyl butyl ester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.