missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl 4-Methoxycinnamate, TRC
Specifications
Specifications
| Chemical Name or Material | Isoamyl Methoxycinnamate |
| CAS | 71617-10-2 |
| Molecular Formula | C15 H20 O3 |
| Quantity | 1 g |
| Synonym | 4-Methoxycinnamic Acid-Isoamyl Ester, 3-Methylbutyl 3-(4-methoxyphenyl)prop-2-enoate, 4-Methoxycinnamic acid 2-isoamyl ester, 4-Methoxycinnamic acid isoamyl ester, 4-Methoxycinnamic acid isopentyl ester, Amiloxate, E 1000, Isoamyl 4-methoxycinnamate, Isoamyl methoxycinnamate, Isoamyl p-methoxycinnamate, Isopentyl 4-methoxycinnamate, Isopentyl p-methoxycinnamate, NSC 408332, Neo Heliopan 1000, Neo Heliopan E 1000 |
| InChI Formula | InChI=1S/C15H20O3/c1-12(2)10-11-18-15(16)9-6-13-4-7-14(17-3)8-5-13/h4-9,12H,10-11H2,1-3H3/b9-6+ |
| SMILES | COc1ccc(\C=C\C(=O)OCCC(C)C)cc1 |
| IUPAC Name | 3-methylbutyl (E)-3-(4-methoxyphenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 248.32 |
| Formula Weight | 248.1412 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?