missing translation for 'onlineSavingsMsg'
Learn More

Isoallolithocholic Acid, TRC

Product Code. 30404084
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404084 100 mg 100mg
30403390 50 mg 50mg
30399685 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404084 Supplier Toronto Research Chemicals Supplier No. TRCI267450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2276-93-9
Molecular Formula C24H40O3
Molecular Weight (g/mol) 376.58
Synonym (3beta,5alpha)-3-Hydroxycholan-24-oic Acid, 3beta-Hydroxy-5alpha-cholanic Acid, 3beta-Hydroxy-5alpha-cholanoic Acid, NSC 18169, (R)-4-((3S,5S,8R,9S,10S,13R,14S,17R)-3-Hydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic Acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16+,17+,18+,19-,20+,21+,23+,24-/m1/s1
Formula Weight 376.2977
Chemical Name or Material Isoallolithocholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.