missing translation for 'onlineSavingsMsg'
Learn More

Isoacteoside, MedChemExpress

Product Code. 16488956
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488956 100 mg 100mg
16468828 10 mg 10mg
16404737 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488956 Supplier Medchem Express Supplier No. HYN0022
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Isoacteoside is a natural compound which exhibit significant inhibition of advanced glycation end product formation with IC50 values of 4.6-25.7 μM, compared with those of aminoguanidine (IC50=1,056 μM) and quercetin (IC50=28.4 μM) as positive controls.

Isoacteoside is a natural compound which exhibit significant inhibition of advanced glycation end product formation with IC50 values of 4.6-25.7 μM, compared with those of aminoguanidine (IC50=1,056 μM) and quercetin (IC50=28.4 μM) as positive controls.

Specifications

Chemical Name or Material Isoacteoside
CAS 61303-13-7
Color White
Purity Grade Notes Research
Molecular Formula C29H36O15
Quantity 100 mg
Synonym Isoverbascoside
Solubility Information DMSO : 250 mg/mL (400.26 mM; Need ultrasonic)
SMILES OC1=CC(/C=C/C(OC[C@@H]2[C@@H](O)[C@H](O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)[C@@H](O)[C@H](OCCC4=CC=C(O)C(O)=C4)O2)=O)=CC=C1O
Molecular Weight (g/mol) 624.59
Formula Weight 624.59
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.