missing translation for 'onlineSavingsMsg'
Learn More

Iso-Valeric Acid, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10365960 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10365960 100 mL Plastic Coated Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10365960 Supplier Fisher Chemical Supplier No. V/0011/PB05
  • ONLINE EXCLUSIVE PRICE
    £281.00
    £276.00 / 100mL
    Save £5.00
    1% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 503-74-2
Melting Point -35°C
pH 3
Boiling Point 176°C
Packaging Plastic Coated Glass bottle
Molecular Formula C5H10O2
MDL Number MFCD00002726
Quantity 100 mL
Synonym isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
InChI Key GWYFCOCPABKNJV-UHFFFAOYSA-N
SMILES CC(C)CC(O)=O
IUPAC Name 3-methylbutanoic acid
Molecular Weight (g/mol) 102.13
PubChem CID 10430
ChEBI CHEBI:28484
Formula Weight 102.13g/mol
Vapor Pressure 0.5 hPa at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Isovaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.