missing translation for 'onlineSavingsMsg'
Learn More

Iso Sildenafil, TRC

Product Code. 30406218
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406218 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406218 Supplier Toronto Research Chemicals Supplier No. TRCI90080010MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 253178-46-0
Molecular Formula C22 H30 N6 O4 S
Quantity 10 mg
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-2,6-dihydro-2-methyl-3-propyl-, Piperazine, 1-[[3-(4,7-dihydro-2-methyl-7-oxo-3-propyl-2H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl- (9CI), 5-[2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-2,6-dihydro-2-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Isosildenafil
InChI Formula InChI=1S/C22H30N6O4S/c1-5-7-17-19-20(25-27(17)4)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29)
SMILES CCCc1c2N=C(NC(=O)c2nn1C)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4
IUPAC Name 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-2-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol) 474.58
Formula Weight 474.2049
Chemical Name or Material Iso Sildenafil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.