missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Safety Sign, Floor Marking - Disinfect Your Hands

Product Code. 30280548
Click to view available options
Languages:
Danish
Dutch
English
Finnish
French
German
Italian
Norwegian
Portuguese
Spanish
Swedish
Turkish
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30280548

Brand: Brady™ 323396

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Floor sign to signify that hands must be disinfected.

  • Sign compliant to ISO 7010

Specifications

Certifications/Compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M061/DE453/FLO-DIA350-1
Grade M061
Material Polyester
Quantity 1 Ea.
Diameter (Metric) 350 mm
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Floor Marking
Thickness 0.28 mm
Shape Round
Languages German
Displayed Text STOPP Hände desinfizieren
Product Type Personal Hygiene Sign
Color White on Blue
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.