missing translation for 'onlineSavingsMsg'
Learn More

Iso Rizatriptan, TRC

Product Code. 30390907
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390907 1 mg -
30388801 10 mg 10mg
30404357 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390907 Supplier Toronto Research Chemicals Supplier No. TRCI8955001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208941-96-2
Molecular Formula C15H19N5
Molecular Weight (g/mol) 269.34
Synonym N,N-Dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-2-ethanamine, Rizatriptan EP Impurity C
IUPAC Name N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-2-yl]ethanamine
SMILES CN(C)CCc1cc2cc(Cn3cncn3)ccc2[nH]1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H19N5/c1-19(2)6-5-14-8-13-7-12(3-4-15(13)18-14)9-20-11-16-10-17-20/h3-4,7-8,10-11,18H,5-6,9H2,1-2H3
Formula Weight 269.164
Chemical Name or Material Iso Rizatriptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.