missing translation for 'onlineSavingsMsg'
Learn More

Iso Ribavirin (Ribavirin Impurity G), TRC

Product Code. 30401868
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401868 1 mg -
30400602 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401868 Supplier Toronto Research Chemicals Supplier No. TRCI8950001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39030-43-8
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.2
Synonym 1-beta-D-Ribofuranosyl-1H-1,2,4-triazole-5-carboxamide, Ribavirin Impurity G
IUPAC Name 2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES NC(=O)c1ncnn1[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H12N4O5/c9-6(16)7-10-2-11-12(7)8-5(15)4(14)3(1-13)17-8/h2-5,8,13-15H,1H2,(H2,9,16)/t3-,4-,5-,8-/m1/s1
Formula Weight 244.0808
Chemical Name or Material Iso Ribavirin (Ribavirin Impurity G)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.