missing translation for 'onlineSavingsMsg'
Learn More

Iso Loratadine, TRC

Product Code. 30011434
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011434 25 mg 25mg
30003226 10 mg 10mg
30003456 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011434 Supplier Toronto Research Chemicals Supplier No. TRCI82105025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170727-59-0
Molecular Formula C22 H23 Cl N2 O2
Molecular Weight (g/mol) 382.88
Synonym Ethyl 4-[(11RS)-8-Chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]-3,6-dihydropyridine-1(2H)-carboxylate, 4-(8-chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl)-3,6-dihydro-1(2H)-pyridinecarboxylic acid ethyl ester, Isoloratadine
IUPAC Name ethyl 4-(8-chloro-6,11-dihydro-5H-benzo[1,2]cyclohepta[2,4-b]pyridin-11-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CCOC(=O)N1CCC(=CC1)C2c3ccc(Cl)cc3CCc4cccnc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-9,11,14,20H,2,5-6,10,12-13H2,1H3
Formula Weight 382.1448
Percent Purity >95%
Chemical Name or Material Iso Loratadine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.