Product Code. 16605913
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
16605913
200 L
200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
16605913
Supplier
Burdick & Jackson BioSyn
Supplier No.
65824200LRC
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Specifications
Melting Point
-154°C
CAS
64742-49-0
Color
Colorless
Density
0.66 g/cm3 (20°C)
Quantity
200 L
Flash Point
-26°C
Linear Formula
CH3 (CH2 )4 CH3
MDL Number
MFCD00081849
Boiling Point
48°C to 65°C (1013 hPa)
IUPAC Name
Petroleum ether
Chemical Name or Material
iso-Hexane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?